Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE/iFluor® 700)

Supplier: AAT Bioquest
100211X0 100211X1 100211X2
77021-830EA 210.7 USD
77021-830 77024-482 77024-974
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE/iFluor® 700)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PE/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR