Stoddard solvent (denatured with 5% ethylacetate and 1% gasoline), Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Mineral spirits

SX0995-1
EM-SX0995-1CS 1361 USD
EM-SX0995-1
Stoddard solvent (denatured with 5% ethylacetate and 1% gasoline), Supelco®
Stoddard solvent

Mixture, ca. C₁₀ hydrocarbons

Boiling Pt: 156…202 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.79 g/cm³ (20 °C)
MDL Number: MFCD00132767
CAS Number: 8052-41-3
UN: 1268
ADR: 3 III

Order Now

Specification Test Results

Boiling range 153-210°C
Color (APHA) 20 max
Form Clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR