Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 560)

Supplier: AAT Bioquest
104410A0 104410A1
77023-182EA 244.24 USD
77023-182 77024-884
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 560)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 560
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR