Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PE/iFluor® 594)

Supplier: AAT BIOQUEST INC
103401Y0 103401Y1 103401Y2
77024-978EA 210.7 USD
77024-978 77022-454 77022-066
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (PE/iFluor® 594)
Antibodies
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR