Anti-CD19 Mouse Monoclonal Antibody (PE/Texas Red®) [clone: HIB19]

Supplier: AAT BIOQUEST INC
101921S0 101921S1 101921S2
77022-388EA 210.7 USD
77022-388 77022-444 77024-136
Anti-CD19 Mouse Monoclonal Antibody (PE/Texas Red®) [clone: HIB19]
Antibodies
HIB19 is an anti-human monoclonal antibody that recognizes the CD19 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE/Texas Red®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR