Quinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Q0011-500G
Q0011-25G
TCQ0011-500GEA
137
USD
TCQ0011-500G
TCQ0011-25G
Quinoline ≥98.0% (by GC, titration analysis)
Quinoline
CAS Number: 91-22-5
MDL Number: MFCD00006736
Molecular Formula: C9H7N
Molecular Weight: 129.16
Purity/Analysis Method:
Formula:
C₉H₇N MW: 129.16 g/mol Boiling Pt: 238 °C Melting Pt: –15 °C Density: 1.10 (20 °C) Flash Pt: 106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III Merck Index: 13,08160 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...