Sterile Nitrile Gloves, Kimtech™ G3, Kimberly-Clark
Supplier: Kimberly-Clark
56888 56894 56887 56893 56889 56890 56891 56892
32933-980CS 807.19 USD
32933-980 41012-992 41012-994 41012-990 41012-982 41012-984 41012-986 41012-988
Sterile Nitrile Gloves, Kimtech™ G3, Kimberly-Clark
Gloves
Gloves are packaged in a ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom and contain no natural rubber latex
  • No natural and rubber latex, powder-free
  • Sterilized by gamma radiation
  • Walleted and pouched in polyethylene for aseptic donning
  • Low endotoxin levels
  • Recommended for ISO Class 4 or higher (FED-STD-209E Class 1/M1.5) cleanroom environments
  • Hand specific, textured palm and fingertips, beaded cuff
Sterilized by gamma radiation. Walleted and pouched in polyethylene for cleanroom use and aseptic donning. Low endotoxin levels. Recommended for ISO Class 4 or higher (FED-STD-209E Class 1/M1.5) cleanroom environments. Hand-specific. Powder-free. Textured palm and fingertips - AQL 1.5. Beaded cuff. Color: white.
Ordering information: Packaged in a Class 100 cleanroom for aseptic donning. Each pair is double-packaged in a wallet within a pouch, 20 pouches per pack.