Water cell culture reagent DNase-, RNase-free

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Dihydrogen oxide

04821739
IC821739EA 123.95 USD
IC821739
Water cell culture reagent DNase-, RNase-free
Water

Sterile water. Deionized, treated with 0.001% DEPC, 0.2µm filtered, and autoclaved. DNase- and RNase-free.


  • DNase-free
  • RNase-free


Deionized water treated with 0.001% diethylpyrocarbonate (DEPC). Filtered through 0.2 micron filter and autoclaved to yield a sterile solution completely free of detectable DNase (both exo and endo) and RNase.


Treatment: Deionized and treated with 0.001% diethylpyrocarbonate (DEPC). It is then filtered through a 0.2 micron filter and autoclaved to yield a sterile solution.Completely Free of DNase and RNase.


Water, DEPC-Treated, is a DEPC(Diethylpyrocarbonate)-treated and sterile filtered water. DEPC inactivates RNase enzymes in water by covalent modifications of the histidine residues. Water, DEPC-Treated is used for the reconstitution of lyophilized products such as oligomers, proteins, and siRNA

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
https://us-prod2.vwr.com/stibo/web/std.lang.all/99/59/36399959.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR