1,2-Dimethoxyethane ≥99.0%

Supplier: TCI America
Danger

Synonyms: Dimethyl glycol, Ethylene glycol dimethyl ether, DME

D0634-500ML D0634-25ML
TCD0634-500MLEA 86.02 USD
TCD0634-500ML TCD0634-25ML
1,2-Dimethoxyethane ≥99.0%
1,2-Dimethoxyethane

CAS Number: 110-71-4 MDL Number: MFCD00008502 Molecular Formula: C4H10O2 Molecular Weight: 90.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 84 Flash Point (°C): -2 Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Dimethyl Cellosolve, Dimethyl Glycol, DME, Ethylene Glycol Dimethyl Ether

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 85
Melting Pt: -69
Density: 0.87 (20 °C)
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008502
CAS Number: 110-71-4
EINECS: 203-794-9
UN: 2252
ADR: 3 II
Merck Index: 13,03251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=774013f8f155aa30ac65be3f869bd25fadc3643e&vid=774013f8f155aa30ac65be3f869bd25fadc3643e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR