Sodium deoxycholate, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
Deoxycholic Acid Sodium Salt Monohydrate is an anionic biological detergent for the solubilization of membrane-bound proteins.
- Aggregation Number: 4-10 (Lit.)
- Critical Micelle Concentration (CMC): 5mM
Anionic detergent; useful for extraction of membrane receptors and other plasma membrane proteins and for nuclei isolation. Deoxycholic acid has been used in a modified procedure to recover 40-80% of a protein from a 1 µg/ml solution.
It solubilizes fats for absorption in the intestines.
Deoxycholic Acid, Sodium Salt is reported to induce tyrosine phosphorylation of b-catenin and its subsequent translocation to the nucleus, promoting activation of β-catenin signaling (effective concentration 5-50 µM). It solubilizes fats for absorption in the intestines.
Soluble in 2% autoclaved solution (clear), and aqueous solutions (50 mg/mL-colorless to light yellow solution).
Caution: Research Use Only (RUO).
Formula:
C₂₄H₃₉NaO₄ MW: 414.56 g/mol Melting Pt: 190 °C (dec., lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00064139 CAS Number: 302-95-4 EINECS: 206-132-7 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
pH (2% aq soln) | 7.5-9.5 |
Loss on Drying |
|
Sodium Cholate | ≤2% |
Purity | Report Result |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...