Xylitol ≥98.0%

Supplier: TCI America

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

X0018-25G X0018-500G
TCX0018-025GEA 17.11 USD
TCX0018-025G TCX0018-500G
Xylitol ≥98.0%
Xylitol

CAS Number: 87-99-0 MDL Number: MFCD00064292 Molecular Formula: C5H12O5 Molecular Weight: 152.15 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Melting point (°C): 92

Formula: C₅H₁₂O₅
Boiling Pt: 495 °C (1013 hPa)
Melting Pt: 92 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 13,10140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d831969c9a136fe5831a70aa90976e72eee0de9f&vid=d831969c9a136fe5831a70aa90976e72eee0de9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR