3-Quinolinecarbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

OR30994-100MG OR30994-50MG
77419-306EA 23.32 USD
77419-306 77419-308
3-Quinolinecarbaldehyde
3-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00006768
CAS Number: 13669-42-6
EINECS: 237-147-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR