Sodium hexafluoroantimonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Sodium hexafluoroantimonate(V)

PC6480-500G PC6480-100G PC6480-25G
77417-414EA 151.8 USD
77417-414 77417-410 77417-412
Sodium hexafluoroantimonate 98%
Sodium hexafluoroantimonate
Formula: F₆NaSb
Melting Pt: 1360 °C
Density: 3.375 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003482
CAS Number: 16925-25-0
EINECS: 240-989-8
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR