1-Methylpiperazine

Supplier: APOLLO SCIENTIFIC
Danger

OR18370-1KG OR18370-250G
77406-302EA 137.54 USD
77406-302 77406-304
1-Methylpiperazine
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138
Melting Pt: -6
Density: 0.903 g/cm³ (25 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR