3-Tolylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Methylphenyl)metaboric acid, Dihydroxy(3-methylphenyl)borane, 3-Methylbenzeneboronic acid, 3-Methylphenylboronic acid, m-Tolylboronic acid

OR0643-100G OR0643-25G OR0643-500G
77402-168EA 80.48 USD
77402-168 77402-170 77402-172
3-Tolylboronic acid 97%
3-Tolylboronic acid
Formula: C₇H₉BO₂
MW: 135.96 g/mol
Melting Pt: 165
Storage Temperature: Refrigerator
MDL Number: MFCD00040198
CAS Number: 17933-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR