Ethylene carbonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR922347-100G OR922347-25G OR922347-500G
77425-056EA 59.1 USD
77425-056 77425-058 77425-060
Ethylene carbonate 98%
Ethylene carbonate
Formula: C₃H₄O₃
Boiling Pt: 243-244/740mm °C
Melting Pt: 35-38 °C
Density: 1.321
Flash Pt: 143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005382
CAS Number: 96-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR