2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(2-Methylbut-3-yn-2-yloxy)oxane

OR70332-5G OR70332-1G OR70332-25G
77424-056EA 73.36 USD
77424-056 77424-052 77424-054
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran
Formula: C₁₀H₁₆O₂
MW: 168.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00090526
CAS Number: 27943-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR