2-Amino-5-cyclopropylbenzoic acid 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR70327-1G
OR70327-250MG
OR70327-5G
77411-006EA
496.16
USD
77411-006
77411-008
77411-010
2-Amino-5-cyclopropylbenzoic acid 95%
2-Amino-5-cyclopropylbenzoic acid
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol |
MDL Number:
MFCD06802840 CAS Number: 68701-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...