4-Aminopyridine 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Pyridin-4-amine, 4-Pyridylamine
OR5929-250G
OR5929-50G
77410-566EA
268.96
USD
77410-566
77410-568
4-Aminopyridine 98%
4-Aminopyridine
Formula:
C₅H₆N₂ Boiling Pt: 273 °C Melting Pt: 155…158 °C |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 UN: 2671 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...