N-Methylanthranilic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Methylamino)benzoic acid

OR10023-1G OR10023-5G
77402-398EA 21.26 USD
77402-398 77402-400
N-Methylanthranilic acid 99%
N-Methylanthranilic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 80
Melting Pt: 172
MDL Number: MFCD00002424
CAS Number: 119-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR