1,3-Dichloropropane 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Trimethylendichloride
OR0973-100G
OR0973-250G
OR0973-25G
77402-356EA
85.58
USD
77402-356
77402-358
77402-360
1,3-Dichloropropane 99%
1,3-Dichloropropane
Formula:
ClH₂CCH₂CH₂Cl MW: 112.99 Boiling Pt: 120.4 Melting Pt: -99.5 Density: 1.19 Flash Pt: 21 °C |
MDL Number:
MFCD00000999 CAS Number: 142-28-9 UN: 1933 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...