2-Chloro-5-(trifluoromethyl)-4-iodopyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

PC9878-1G PC9878-25G
77434-520EA 21.34 USD
77434-520 77434-522
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Formula: C₆H₂ClF₃IN
Melting Pt: 126…127 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR