1-Bromoperfluorohexane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluorohexyl bromide

PC6085W-100G PC6085W-25G
77432-480EA 381.04 USD
77432-480 77432-482
1-Bromoperfluorohexane 98%
1-Bromoperfluorohexane
Formula: BrF₂C(CF₂)₄CF₃
MW: 398.95 g/mol
Boiling Pt: 97…101 °C
Density: 1.871 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042349
CAS Number: 335-56-8
EINECS: 206-391-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR