5-Isoquinolineboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR3296-100MG OR3296-1G
77420-190EA 30.56 USD
77420-190 77420-192
5-Isoquinolineboronic acid 97%
Formula: C₉H₈BNO₂
Melting Pt: 186…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839356
CAS Number: 371766-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR