4-Bromo-3-(trifluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-5-(bromomethyl)benzotrifluoride, 1-Bromo-4-(bromomethyl)-2-(trifluoromethyl)benzene

PC6025-1G PC6025-250MG PC6025-5G
77417-262EA 99.84 USD
77417-262 77417-264 77417-266
4-Bromo-3-(trifluoromethyl)benzyl bromide
4-Bromo-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₅Br₂F₃
MW: 317.929 g/mol
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD08059128
CAS Number: 1159512-68-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR