Hexakis(3,3,3-trifluoropropoxy)phosphazene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC450559-1G PC450559-250MG
77429-960EA 545.06 USD
77429-960 77429-962
Hexakis(3,3,3-trifluoropropoxy)phosphazene 95%
Hexakis(3,3,3-trifluoropropoxy)phosphazene
Formula: C₁₈H₂₄F₁₈N₃O₆P₃
MW: 813.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075243
CAS Number: 1980062-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR