N-((Dimethylamino)methylene)-2,4-difluorobenzamide

Supplier: APOLLO SCIENTIFIC
Warning

PC28281-1G PC28281-250MG
77427-894EA 93.74 USD
77427-894 77427-896
N-((Dimethylamino)methylene)-2,4-difluorobenzamide
N-((Dimethylamino)methylene)-2,4-difluorobenzamide
Formula: C₁₀H₁₀F₂N₂O
MW: 212.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812108
CAS Number: 2379917-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR