Aminotriester 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Di-tert-butyl 4-amino-4-(3-(tert-butoxy)-3-oxopropyl)heptanedioate

OR931286-1G OR931286-250MG OR931286-5G
77425-312EA 171.16 USD
77425-312 77425-314 77425-316
Aminotriester 95%
Aminotriester
Formula: C₂₂H₄₁NO₆
MW: 415.564 g/mol
Melting Pt: 60…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07357283
CAS Number: 136586-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR