2-Amino-4-bromo-6-chlorophenol

Supplier: APOLLO SCIENTIFIC

OR400332-1G OR400332-250MG OR400332-5G
77420-878EA 73.36 USD
77420-878 77420-880 77420-882
2-Amino-4-bromo-6-chlorophenol
2-Amino-4-bromo-6-chlorophenol
Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16742724
CAS Number: 855836-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR