3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: Antioxidant 1024, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)propanehydrazide, 1,2-Bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamoyl)hydrazine

OR933276-25G OR933276-5G
77425-384EA 116.14 USD
77425-384 77425-386
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 95%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide
Formula: C₃₄H₅₂N₂O₄
MW: 552.80 g/mol
Boiling Pt: 60…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072662
CAS Number: 32687-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR