4-Ethynylbenzyl alcohol 99%

Supplier: APOLLO SCIENTIFIC

Synonyms: (4-Ethynylphenyl)methanol

OR939454-5G OR939454-10G OR939454-1G
77425-516EA 101.88 USD
77425-516 77425-512 77425-514
4-Ethynylbenzyl alcohol 99%
4-Ethynylbenzyl alcohol
Formula: C₉H₈O
MW: 132.16 g/mol
Melting Pt: 40…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664205
CAS Number: 10602-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR