Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

PC10246-100MG PC10246-250MG
77426-554EA 39.32 USD
77426-554 77426-556
Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate
Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₀FIN₂O₂
MW: 360.13 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD03791218
CAS Number: 959576-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR