3-(Trifluoromethoxy)-DL-phenylglycine

Supplier: APOLLO SCIENTIFIC
Warning

PC0517-1G PC0517-250MG
77426-260EA 182.36 USD
77426-260 77426-262
3-(Trifluoromethoxy)-DL-phenylglycine
3-(Trifluoromethoxy)-DL-phenylglycine
Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631460
CAS Number: 261952-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR