Boc-Aib-ol 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-Boc-2-amino-2-methyl-1-propanol, N-tert-Butoxycarbonyl-2-amino-2-methyl-1-propanol, tert-Butyl (1-Hydroxy-2-methylpropan-2-yl)carbamate

OR931504-1G OR931504-25G OR931504-5G
77425-318EA 21.26 USD
77425-318 77425-320 77425-322
Boc-Aib-ol 96%
Boc-Aib-ol
Formula: C₉H₁₉NO₃
MW: 189.3 g/mol
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03788641
CAS Number: 102520-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR