1H-Indole-2-carbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Indole-2-carbonitrile

OR310037-250MG OR310037-1G
77419-316EA 55.02 USD
77419-316 77419-314
1H-Indole-2-carbonitrile
1H-Indole-2-carbonitrile
Formula: C₉H₆N₂
MW: 142.161 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00187566
CAS Number: 36193-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR