Sodium trifluoromethanesulfinate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent, Trifluoromethanesulfinic acid Sodium Salt

PC0579-100G PC0579-25G PC0579-5G
77426-286EA 49.92 USD
77426-286 77426-288 77426-290
Sodium trifluoromethanesulfinate 95%
Sodium trifluoromethanesulfinate
Formula: CHF₃O₂S
MW: 156.05 g/mol
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD03092989
CAS Number: 2926-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR