Octafluoroadipic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Perfluoroadipic Acid, Octafluorohexanedioic Acid, Perfluorohexanedioic Acid

PC5320-1G PC5320-250MG PC5320-5G
77431-386EA 28.52 USD
77431-386 77431-388 77431-390
Octafluoroadipic acid 97%
Octafluoroadipic acid
Formula: C₆H₂F₈O₄
Boiling Pt: 165 °C/1 mmHg
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00066405
CAS Number: 336-08-3
EINECS: 206-407-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR