N-Decanoylsarcosine sodium salt 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: Sodium N-Decanoylsarcosinate, (N-Decanoyl-N-methylamino)acetic Acid Sodium Salt, Sodium (N-Decanoyl-N-methylamino)acetate

OR936414-1G OR936414-5G
77425-408EA 166.06 USD
77425-408 77425-410
N-Decanoylsarcosine sodium salt 98%
N-Decanoylsarcosine sodium salt
Formula: C₁₃H₂₅NO₃
MW: 265.33 g/mol
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00451533
CAS Number: 30377-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR