1-Methyl-1H-pyrazole-4-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR54372-1G OR54372-250MG OR54372-5G
77410-374EA 24.46 USD
77410-374 77410-376 77410-378
1-Methyl-1H-pyrazole-4-sulfonyl chloride
1-Methyl-1H-pyrazole-4-sulfonyl chloride
Formula: C₄H₅ClN₂O₂S
MW: 179.98 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Freezer
MDL Number: MFCD04968667
CAS Number: 288148-34-5
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR