(S)-(+)-Camptothecin

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

OR014824-1G OR014824-5G
77401-704EA 27.5 USD
77401-704 77401-706
(S)-(+)-Camptothecin
(S)-(+)-Camptothecin
Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00081076
CAS Number: 7689-03-4
UN: 1544
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR