2-(Thiazol-2-yl)benzaldehyde
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR1995-5G
OR1995-1G
77406-514EA
303.6
USD
77406-514
77406-512
2-(Thiazol-2-yl)benzaldehyde
2-(Thiazol-2-yl)benzaldehyde
Formula:
C₁₀H₇NOS MW: 189.2300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06657778 CAS Number: 223575-69-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...