1-Pyreneboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR40378-10G OR40378-5G
77421-116EA 59.1 USD
77421-116 77421-118
1-Pyreneboronic acid 97%
1-Pyreneboronic acid
Formula: C₁₆H₁₁BO₂
Melting Pt: 247…251 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974062
CAS Number: 164461-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR