1-Boc-piperidin-4-ylboronic acid pinacol ester 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, Piperidine-4-boronic acid, pinacol ester, N-BOC protected

OR30822-10G OR30822-1G OR30822-5G
77419-192EA 221.08 USD
77419-192 77419-194 77419-196
1-Boc-piperidin-4-ylboronic acid pinacol ester 96%
1-Boc-piperidin-4-ylboronic acid pinacol ester
Formula: C₁₆H₃₀BNO₄
MW: 311.2247 g/mol
Melting Pt: 60…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09997792
CAS Number: 1048970-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR