2-(Trifluoromethyl)-4-quinolinol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline

PC5632-1G PC5632-5G
77416-834EA 21.26 USD
77416-834 77416-836
2-(Trifluoromethyl)-4-quinolinol 95+%
2-(Trifluoromethyl)-4-quinolinol
Formula: C₁₀H₆F₃NO
MW: 213.1560 g/mol
Melting Pt: 198…203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00153196
CAS Number: 1701-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR