(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (R)-4-Hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide, (R)-(-)-1,1'-Binaphthalene-2,2'-diyl-phosphate

OR315004-25G OR315004-5G
77419-504EA 109.02 USD
77419-504 77419-506
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Formula: C₂₀H₁₃O₄P
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS Number: 39648-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR