6-Aminoimidazo[1,2-a]pyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Imidazo[1,2-a]pyridin-6-amine

OR305164-5G OR305164-100MG OR305164-1G
77418-916EA 207.84 USD
77418-916 77418-912 77418-914
6-Aminoimidazo[1,2-a]pyridine 95%
6-Aminoimidazo[1,2-a]pyridine
Formula: C₇H₇N₃
MW: 133.1506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739232
CAS Number: 235106-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR