2-Chloro-4,5-difluoroaniline

Supplier: APOLLO SCIENTIFIC
Warning

PC302442-100G PC302442-25G PC302442-5G
77428-292EA 158.38 USD
77428-292 77428-294 77428-296
2-Chloro-4,5-difluoroaniline
2-Chloro-4,5-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08448325
CAS Number: 2613-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR