Diethyl aminomalonate hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diethyl 2-aminomalonate hydrochloride

OR28630-100G OR28630-1G OR28630-500G
77408-136EA 41.78 USD
77408-136 77408-138 77408-140
Diethyl aminomalonate hydrochloride 95%
Diethyl aminomalonate hydrochloride
Formula: C₇H₁₄ClNO₄
MW: 211.65 g/mol
Melting Pt: 165…170 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012510
CAS Number: 13433-00-6
EINECS: 236-556-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR