Trithiocyanuric acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11248-100G OR11248-500G
77403-846EA 55.02 USD
77403-846 77403-848
Trithiocyanuric acid 98%
Trithiocyanuric acid
Formula: C₃H₃N₃S₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006052
CAS Number: 638-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR