Hydroxylamine-O-sulfonic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Amino hydrogen sulfate, Amino hydrogen sulphate

OR0911-100G OR0911-1KG OR0911-250G
77402-296EA 48.9 USD
77402-296 77402-298 77402-300
Hydroxylamine-O-sulfonic acid 98%
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MW: 113.09 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR